Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Reduction from sleeve to core.
98.23 DKK - 1535.00 DKK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
10223881
|
DWK Life Sciences
286548201 |
Socket NS 29/32, Cone NS 24/29 | N/A | ||||||
15884818
|
DWK Life Sciences
286548004 |
Socket NS 29/32, Cone NS 14/23 |
1245.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10305101
|
DWK Life Sciences
286548107 |
Socket NS 29/32, Cone NS 19/26 | N/A | ||||||
15981601
|
DWK Life Sciences
286542704 |
Socket NS 24/29, Cone NS 19/26 |
1535.00 DKK
Pack of 10 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
15961601
|
DWK Life Sciences
286535906 |
Socket NS 19/26, Cone NS 14/23 |
1300.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15844818
|
DWK Life Sciences
286542507 |
Socket NS 24/29, Cone NS 14/23 |
1500.00 DKK
Pack of 10 |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
Description
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
- Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
Specifications
10/Pk. |