Learn More
Smeg™ Washer Rack
Designed for use with Smeg™ glassware washing devices. Smeg Washer Racks can be used as racks to hold small glass items or as an alternative to the Nylon holding nets.
Marca:  Smeg™ PF2
Codice prodotto. 11945624
  • 254.00€ / Pezzo
Solo null disponibili
Spedizione stimata: 25-06-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Additional accessories for the small glassware baskets CS1-1 and CS2
  • Flat AISI 304 stainless steel mesh with small mesh openings 
  • PF1 bottom rack with hole for positioning on the top level
  • PF2 complete bottom rack for positioning on the bottom level only
  • PF1/2 half bottom rack for positioning on the top level only
Specifica
Specifica
Stainless Steel
Rack
Complete bottom rack for positioning on the bottom level only
Small glassware, baskets and holding nets