Learn More
Sampling Systems™ TeleSwab™ Telescopic Swabbing Tool Complete Kits
The TeleSwab™ Telescopic Swabbing Tool allows operators to quickly and easily take swabs from previously inaccessible places.
Marca: Sampling Systems™ 8011K-01
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Ideal for environmental swabbing and for use with large items of processing equipment
- Extendable handle that can be extended up to 3m
- Adjustable adapter for angling swabs to meet the contours of the item being sampled
- PTFE template tool allows a fixed area to be swabbed
- Two different complete kits are available
- Kit 1 contains one telescopic pole, one angle adapter and one swab holder
- Kit 2 contains one telescopic pole, one template holder and one swabbing template with a hole cut to customer’s requirements
Specifica
Swabbing Kit | |
Telescoping | |
Environmental swabbing |
Angle Adapter, Swab Holder | |
Kit 1 | |
Extendable to 3 m |