Learn More
Cole-Parmer™ Tube Holder
Tube Holder for Use with Microtitre Plate Shakers SSM5 and SSL5.
166.00 EUR - 173.00 EUR
Specifications
Product Type | Tube Holder |
---|---|
For Use With (Equipment) | Stuart™ Microtitre Plate Shakers SSM5 and SSL5 |
Packaging Quantity | 1 Ea. |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12693335
|
Cole-Parmer™
51900-99 |
0.2 mLTubes |
166.00 EUR
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
12944025
|
Cole-Parmer™
51900-98 |
0.5 mL Tubes |
173.00 EUR
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
11496608
|
Cole-Parmer™
51900-97 |
1.5 mL Tubes |
172.00 EUR
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
Description
- Accessory tube holders accommodate 0.2 mL or 0.5 mL microcentrifuge tubes.
- Two tube holders can be used at any one time and securely held in place by the non-slip mat.
- Stuart™ Tube Holder; For Use with Microtitre Plate Shakers where samples require controlled temperature and/or humidity conditions, ideal for placement inside CO2incubators.
Specifications
Tube Holder | |
Stuart™ Microtitre Plate Shakers SSM5 and SSL5 | |
1 Ea. |