Learn More
Afora™ Short Neck Round Flask with Flange
Boil laboratory chemicals using the Afora™ Short Neck Round Flask with Flange. Its quality construction makes it durable and reliable in the laboratory.
28.00€ - 103.00€
Specifications
Description | Top style: With flange |
---|---|
Material | Borosilicate Glass |
Bottom Shape | Round |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11921588
|
Afora™
2 |
73 mm |
28.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
12339910
|
Afora™
3/14 |
88 mm | N/A | ||||||
11932258
|
Afora™
4/14 |
103 mm | N/A | ||||||
11922268
|
Afora™
4/29 |
113 mm | N/A | ||||||
12023898
|
Afora™
7 |
133 mm |
77.00€
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
12060362
|
Afora™
8/29 |
152 mm |
87.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
12040592
|
Afora™
9 |
182 mm |
103.00€
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
Description
- With flange
- Borosilicate glass
- Spherical bottom
- Shorter neck
Specifications
Top style: With flange | |
Borosilicate Glass | |
Round |