Learn More
Afora™ Short Neck Round Flask with Flange
Boil laboratory chemicals using the Afora™ Short Neck Round Flask with Flange. Its quality construction makes it durable and reliable in the laboratory.
Brand: Afora™ 8/29
10.61 EUR valid until 2025-07-22
Use promo code "24098" to get your promotional price.
This item is not returnable.
View return policy
Description
- With flange
- Borosilicate glass
- Spherical bottom
- Shorter neck
Specifications
Top style: With flange | |
Borosilicate Glass | |
Single |
500 mL | |
152 mm | |
Round |