Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Component of the system for the safe loosening of stuck ground joints.
Marca: DWK Life Sciences 286216606
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
- By means of an even force distribution, stuck joints can be safely loosened.
- Due to their material properties, pull rings have very good sliding properties.
Specifica
10/Pk. | |
To secure the security grinding | |
Safeguard | |
NS 29/32 |