Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
Marca: Bohlender™ H912-06
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Precast tweezers
- PTFE construction
- With pointed or blunt ends
- Universal chemical resistance
Specifica
Blunt | |
Precast tweezer | |
200 mm |
Each | |
PTFE | |
Tweezers |