Learn More
Masterflex™ Raccordo scanalato per connettore riduttore in polietilene
Combine these simple, economical HDPE reducing connector with flexible tubing for use in your low-pressure applications (generally less than 30 psi). HDPE Reducing Connector Barbed Fittings slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
Marca: Masterflex™ 30622-03
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Designed for quick and easy assembly without tools
- Very Good chemical resistance
- Temp range: - 53 to 65°C
- Sterilize by ethylene oxide
Specifica
Polyethylene | |
5/8 x 1/2 in. tubing I.D. | |
10 Pack | |
Barbed Fitting |