Learn More
Thermo Scientific™ Oxoid™ GMO-Free Soya Peptone
Oxoid GMO-Free Soya Peptone is a nutritious, general-purpose peptone for the growth of bacteria and fungi.
Marca: Thermo Scientific™ VG0300T
400.73 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
Oxoid GMO-Free Soya Peptone is a nutritious, general-purpose peptone for the growth of bacteria and fungi.
A nutritious, general purpose peptone for the growth of bacteria and fungi.
Specifica
GMO-Free Soya Peptone | |
Powder | |
Peptone | |
5 kg |
Per uso esclusivo in laboratorio