Learn More
Orai1-Alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 14443-1-AP-20UL
Codice prodotto. 16853084
  • 145.00€ / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
This antibody may be used to block the ion currents mediated by Orai1. ORAI1 contains 2isoforms produced byalternative initiation. 14443-1-AP only recognizes alpha isoform.
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.
Specifica
Specifica
Orai1-Alpha
Polyclonal
Unconjugated
ORAI1
ORAI1
Peptide
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5M848, Q8BWG9, Q96D31
Rabbit
Antigen Affinity Chromatography
RUO
109305, 304496, 84876
-20°C