Learn More
Macherey-Nagel™ Tappi a capsula in alluminio N 11
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
Marca: Macherey-Nagel™ 702995
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Ready assembled crimp closures and plain crimp caps N 11
- Septa are assembled, caps are ready-to-use.
- Septa are shown for illustration purposes only, images upside down.
- Packs of 100 closures each
Specifica
Red/White/Red | |
Argento | |
PTFE rosso/Silicone bianco/PTFE rosso |
8 mm | |
Alluminio, PTFE, Silicone | |
1 mm |