missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LILRB4/CD85k/ILT-3 ELISA Kit
ELISA
712.00€ - 2860.00€
Specifications
Accession Number | CD85 antigen-like family member K; CD85k; glycoprotein 49 B; glycoprotein 49 b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; Mast cell surface glycoprotein Gp49B; monocyte inhibitory receptor HM18 |
---|---|
Assay Range | 1.3 to 300 ng/mL |
Assay Sensitivity | 1.3 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17165069
|
Invitrogen™
EH300RBX5 |
5 x 96 Tests |
2860.00€
Each |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17184593
|
Invitrogen™
EH300RB |
96 Tests |
712.00€
Pack of 96 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
Human LILRB4/CD85k/ILT-3 quantitates human LILRB4/CD85k/ILT-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LILRB4/CD85k/ILT-3.
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].Specifications
CD85 antigen-like family member K; CD85k; glycoprotein 49 B; glycoprotein 49 b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; Mast cell surface glycoprotein Gp49B; monocyte inhibitory receptor HM18 | |
1.3 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
LILRB4 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
LILRB4 (CD85k) | |
4 hrs 45 mins |
1.3 to 300 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
11006 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title