Learn More
Invitrogen™ Human LILRB4/CD85k/ILT-3 ELISA Kit
ELISA
Brand:  Invitrogen™ EH300RB
 View more versions of this product
Product Code. 17184593
  • 712.00€ / Pack of 96
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
This item is not returnable. View return policy
Description
Description
Human LILRB4/CD85k/ILT-3 quantitates human LILRB4/CD85k/ILT-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LILRB4/CD85k/ILT-3.
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD85 antigen-like family member K; CD85k; glycoprotein 49 B; glycoprotein 49 b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; Mast cell surface glycoprotein Gp49B; monocyte inhibitory receptor HM18
1.3 ng/mL
ELISA
Colorimetric Microplate Reader
LILRB4
<10
HRP
RUO
2°C to 8°C
1 hrs 20 mins
1.3 to 300 ng/mL
HRP
Plasma, Serum, Supernatant
11006
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
LILRB4 (CD85k)
4 hrs 45 mins
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.