Learn More
Lenz Laborglasintrumente Cones with Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 01100385
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Acc. to DIN 12249 form G and in addition to standard
- Conical ground cones
- Made of DURAN™ tubing
Specifica
1 Ea. | |
3.5 mm | |
Borosilicate Glass (3.3) |
NS 85/55 | |
80 mm |
Titolo del prodotto
Lenz Laborglasintrumente Cones with Tip > NS 85/55