Learn More
Cytiva Cassette
For use with the Fraction collector F9-C and the ÄKTA avant chromatography system. Cytiva Cassettes are available for Deep-Well Plates or 3mL Tubes for large-scale processing of samples in chromatography applications. Pack of 2.
Marca: Cytiva 28954212
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Cytiva Cassettes are available in packs of two (2) for large-scale processing of samples in chromatography applications using the Fraction collector F9-C and the ÄKTA avant chromatography system.- For Deep-Well Plates (in Flexible fraction collector): Holds 24, 48 or 96 deep-well plate
- For 3mL Tubes: Holds up to 40 tubes
Specifica
Chromatography Applications | |
Cassette | |
For Use With 24, 48 or 96 deep-well plate |
Fraction Collector F9-C and ÄKTA Avant Chromatography System | |
2/Pk. |
Titolo del prodotto
Cytiva Cassette > For Use With: 24, 48 or 96 deep-well plate