Learn More
Bohlender™ BOLA™ Sleeves with Ribs
Sleeves with ribs for vacuum up to 0.1 mbar.
Marca:  Bohlender™ H932-06
Codice prodotto. 17177394
  • 18.00€ / pezzi 5
Solo null disponibili
Spedizione stimata: 04-07-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Sealing rings ('ribs') on the outside
  • Without gripping ring and therefore usable with joint clamps
  • For vacuum up to 0.1 mbar
  • Solid construction for continued use
  • Sealing rings are easily accessible and easy to clean
Specifica
Specifica
Vacuum Up to 0.1 mbar
PTFE
Sleeves with Ribs
US 24/40