Learn More
Bohlender™ BOLA™ Sleeves with Ribs
Sleeves with ribs for vacuum up to 0.1 mbar.
Marca: Bohlender™ H930-09
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sealing rings ('ribs') on the outside
- Without gripping ring and therefore usable with joint clamps
- For vacuum up to 0.1 mbar
- Solid construction for continued use
- Sealing rings are easily accessible and easy to clean
Specifica
Vacuum Up to 0.1 mbar | |
PTFE | |
Sleeves with Ribs | |
NS 40/38 |