Learn More
Miele Professional™ Accessory Booster Pump
Rec in/out water inlet and drain for PLW 8636
Marca: Miele Professional™ 12426490
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Rec in/out water inlet and drain for PLW 8636
- Required in order to pump a medium out of a buffer container
- For use with an existing on-site recycling tank
- Can be used with existing on-site container, e.g. IBC tank
- Installed by Customer Service
Specifica
Booster Pump | |
Rec in | |
Washer-Disinfectors | |
1 Ea. |