Learn More
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
24.64€ - 49.85€
Product Code | Brand | Model | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Price | Quantity & Availability | ||||
15328608
|
Bel-Art™
66000-0038 |
OFV | Valve |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
15318608
|
Bel-Art™
66000-0037 |
OF-30 | Septum | N/A | |||||
15398858
|
Bel-Art™
66306-0571 |
507-OF-7 | NMR sample tube | N/A | |||||
Description
- The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
- Omni-Fit Sample tubes (507-OF-7) sold separately
- Omni-Fit valve (OFV) is a complete accessory set
- Additional septum (OF-30) are available in packages of 6
Specifications
5.0 mm |