Learn More
Thermo Scientific™ I-Chem™ Wide-Mouth Amber Glass Packer with Closure
GREENER_CHOICE
Codice prodotto. 17634855
Click to view available options
Capacità (sistema metrico):
60 mL
120 mL
250 mL
500 mL
950 mL
1250 mL
2500 mL
Livello:
Certified
Processed
Unprocessed
Dimensione chiusura:
33-400 mm
38-400 mm
45-400 mm
53-400 mm
70-400 mm
N. per custodia:
4
6
12
24
missing translation for 'unitSize'
pezzi 12
pezzi 24
pezzi 4
pezzi 6
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with these wide-mouth, amber glass packers.
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with Thermo Scientific™ I-Chem™ Wide-Mouth Amber Glass Packers. Packers are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified jars but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Amber Type III soda-lime glass jar; recommended for light-sensitive samples
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 60 to 2500mL
  • Closures assembled on bottles
Certified jars come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each jar individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed jars are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded, and CofA is not included.
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
TRUSTED_SUSTAINABILITY
Specifications
Capacità 16 oz., 500 mL
Capacità (sistema metrico) 500 mL
Capacità (sistema imperiale) 16 oz.
Parametri di certificazione Semi-VOA, pesticides, PCBs, metals
Colore Amber
Descrizione Amber Glass Packer
Diametro (sistema imperiale) interno 1.63 in.
Diametro (sistema imperiale) 3.18 in.
Diametro (sistema metrico) interno 4 cm
Materiale Glass (Type 3)
N. per custodia 12
Chiusura parte sostitutiva B167-5300BL
Colore chiusura Bianco
Livello Processed
Materiale rivestimento interno PTFE
Volume (sistema imperiale) margine 18 oz.
Volume (sistema metrico) margine 533 mL
Certificazione/Conformità Certified and Processed jars meet or exceed EPA performance based standards for semi-volatile organics, pesticides, PCB's and metals analysis; manufactured under an ISO certified quality management system.
Bocca Larga
Altezza (sistema metrico) 146 mm
Product Type Amber Packer Bottle
Altezza (sistema imperiale) 5.74 in.
Materiale chiusura Polypropylene
Dimensione chiusura 53-400 mm
Diametro (sistema metrico) 81 mm
Tipo di chiusura Closed-top Cap