Learn More
3M™ Welding Visors
Codice prodotto. 11721363
Click to view available options
Product Type:
Welding Visor Headtop
Specifica
Product Type Welding Visor Headtop
Da utilizzare con (applicazione) Molten metal splashes, gas, vapor, and particulates, welding glare, metal fumes, in automotive, welding, and metal fabrication industries
Include 90 x 110 mm filter holder able to hold fixed shade or auto-darkening filters