Learn More
Unity Lab Services™ Premium Pipette Support Plan
Fast, quality-minded service that meets your diverse needs
Brand: Unity Lab Services™ GXP-3X10-DMC
Description
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
- Service for any pipette brand and model
- Reliable service options by our highly trained technicians
- Fast and convenient prepaid pipette shipping packages
Premium – GxP Preventive Maintenance and Calibration
The GxP Preventive Maintenance and Calibration Plan provides a comprehensive service that includes gravimetric evaluation of your pipette's performance prior to any inspection, preventive maintenance or minor repairs. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
- As-received data
- Pipette inspection, liquid leak test, routine preventive maintenance and minor repairs
- Calibration
- Work in a highly regulated environment
- Reduced OOTs (Out of Tolerance) that could invalidate research
† Unity Lab Services™ provides a detailed 15-point pipette inspection of the following single and multi-channel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally:
Checked routinely: Pipette body and display, tip ejection and ejector mechanism, tip cone(s), piston(s) assembly, o-ring(s), springs
Checked as applicable: Plunger bottom, plunger rod, volume lock, shaft coupling nut, seal(s), lubrication, battery recharging, electronics functionality, motor movement
Specifications
Multichannel Pipette (Any Brand and Model) | |
GxP Preventive Maintenance and Calibration, 3 x 10 | |
Gravimetric evaluation of pipettes performance prior to any inspection, preventive maintenance or minor repairs; includes calibration certificate and sticker |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.