missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals

CAS: 75-50-3 | C3H9N | 59.11 g/mol

Identificatori chimici

CAS 75-50-3
Formula molecolare C3H9N
Molecular Weight (g/mol) 59.11
Numero MDL MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Sinonimo trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name N,N-dimethylmethanamine
SMILES CN(C)C
Visualizza altre specifiche

Descrizione

Descrizione

Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

Solubility
Insoluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, brass, zinc, magnesium, aluminum, mercury, mercury oxides, acid chlorides, acid anhydrides, strong reducing agents.
Specifica

Identificatori chimici

75-50-3
59.11
GETQZCLCWQTVFV-UHFFFAOYSA-N
1146
N,N-dimethylmethanamine
C3H9N
MFCD00008327
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
CHEBI:18139
CN(C)C

Specifica

Trimethylamine
0.75
C3H9N
1.3635
UN2924
Air sensitive; Hygroscopic
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
GETQZCLCWQTVFV-UHFFFAOYSA-N
N,N-dimethylmethanamine
1146
59.11
75-50-3
−20°C (−4°F)
(CH3)3N
MFCD00008327
956566
14,9710
Insoluble in water.
CN(C)C
59.11
CHEBI:18139
33% w/w in ethanol
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

GHS H Statement
H225-H314-H302-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H304-H314-H335-H336

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 200-875-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Identificativo prodotto
  • Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane

Avvertenza
  • Pericolo

Categoria di pericolo
  • Tossicità acuta Categoria 4
  • PERICOLO A LUNGO TERMINE PER L'AMBIENTE ACQUATICO Cronico 3
  • Tossicità in caso di aspirazione Categoria 1
  • Lesione oculare grave/irritazione oculare Categoria 1
  • Liquido infiammabile Categoria 2
  • Corrosione/irritazione della pelle Categoria 1B
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3

Indicazioni di pericolo
  • H225-Liquido e vapori facilmente infiammabili.
  • H302-Nocivo se ingerito.
  • H304-Può essere letale in caso di ingestione e di penetrazione nelle vie respiratorie.
  • H314-Provoca gravi ustioni cutanee e gravi lesioni oculari.
  • H318-Provoca gravi lesioni oculari.
  • H332-Nocivo se inalato.
  • H336-Può provocare sonnolenza o vertigini.
  • H412-Nocivo per gli organismi acquatici con effetti di lunga durata.

Consigli di prudenza
  • P210-Tenere lontano da fonti di calore, superfici riscaldate, scintille, fiamme e altre fonti di innesco. Vietato fumare.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P301+P330+P331-IN CASO DI INGESTIONE: sciacquare la bocca. NON provocare il vomito.
  • P303+P361+P353-IN CASO DI CONTATTO CON LA PELLE (o con i capelli): togliere immediatamente tutti gli indumenti contaminati. Sciacquare la pelle/fare una doccia.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P310-Contattare immediatamente un CENTRO ANTIVELENI/un medico .

Informazioni supplementari
  • MIXTURE LIST-Contiene: ethanol, trimethylamine, cyclohexane

SDS
Documenti

Documenti

Product Certifications

For Manufacturing and Laboratory Use. Not for beverage use

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato