Learn More
Thiocyanate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
Codice prodotto. 12912215 Sfoglia Tutto Fisher Chemical Prodotti
100 mL
Click to view available options
Quantità:
100 mL
250 mL
500 mL
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifica
Quantità 100 mL
La soluzione contiene H2O
Concentrazione 1 g/L
Titolo del prodotto
Thiocyanate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ >