Learn More
Thermo Scientific™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™
Perform testing of media, stains, reagents, and identification kits, and evaluate bacteriological procedures with Thermo Scientific ™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™.
Marca: Thermo Scientific™ R4601226
202.85 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
Perform testing of media, stains, reagents, and identification kits, and evaluate bacteriological procedures with Thermo Scientific ™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™.
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Specifica
Parabacteroides distasonis | |
8503™* | |
Culti-Loops Quality Control Organism |
Culti-Loops are CE marked.