Learn More
Thermo Scientific™ BetaMax Base Drop-In Guard Cartridges
Offer convenience, economy and effective protection for extending column lifetime
Brand:  Thermo Scientific™ 5825/3
 View more versions of this product
Product Code. 11987148
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »
Requires : Thermo Scientific Uniguard™ direct-connection or stand-alone holder, sold separately.
Description
Description
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fits Thermo Scientific Uniguard™ direct-connection and stand-alone holders
Features of BetaMax Basic Solid Phase
  • Designed for basic analytes
  • Ultra-high surface coverage for maximum retention
  • Excellent stability for long column lifetimes
Specifications
Specifications
5 μm
10 mm
2.1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions