Learn More
Texwipe™ AlphaMop™
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
156.00€
Codice del prodotto | Marca | Materiale | Product Type | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Materiale | Product Type | Prezzo | Quantità e disponibilità | ||||
11591214
|
Texwipe™
TX7108 |
Polyester | Cleanroom Mop |
156.00€
Pezzo |
Spedizione stimata: 13-10-2025
Eseguire il login per visualizzare lo stock disponibile |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
|||
Descrizione
Specifically designed for the needs of ultraclean production environments. Easily changeable, laundered polyester mop covers fit over a flat, foam-padded head assembly. Lightweight, durable fiberglass handle provides excellent control over swivel head and accesses hard-to-reach areas with ease.
- Enables rapid change of wiping surfaces for adherence to strict wiping protocol ISO Class 4 cleanroom-laundered Alpha™ polyester mop cover
- Replaceable foam pad ensures that the polyester mop cover conforms to textured floors and other surfaces
- Maintains ultralow generation of particles and extractables Resists corrosion, abrasion and commonly used cleaning and disinfecting chemicals such as bleaches, phenols and quaternary ammonium compound
- AlphaMop polyester replacement mopheads and foam pads available separately (150 heads and six pads per case)
- AlphaMop mop covers presaturated with 6% IPA Honeycomb™ 10 double-knit polyester mop covers have an abrasive texture (supplied 150 covers per case)
Specifica
Non-sterile |