Learn More
Fritsch™ Test Sieve
Brand:  Fritsch™ 35.2100.03
Product Code. 10545472
  • POA / Each
Estimated Shipment: 11-07-2025
Explore more special offers
Add to basket
Description
Description
Specifications
Specifications
1 Ea.
Test Sieve
50 mm
200 mm
Stainless Steel
3.35 mm
ASTM - E-11-1995
No. 6
6 Mesh
6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers