Learn More
Sampling Systems™ SteriWare™ Scraper
Designed to be ergonomic, which means they are easy to use and ideal for scraping and cleaning. Sampling Systems™ SteriWare™ Scraper is ideal for use with foodstuffs, cosmetics and pharmaceuticals.
Marca: Sampling Systems™ 8785H-110
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Available in 2 sizes
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contaminaton
- No cleaning required
Specifica
Disposable Scraper | |
Individually bagged | |
Natural | |
110 mm |
HDPE | |
Non-sterile | |
225 mm |