Learn More
Sampling Systems™ SteriWare™ Disposable Powder Probe
Sampling Systems™ SteriWare™ Disposable Powder Probe is ideal for taking a cross sectional sample through a bulk powder.
Marca: Sampling Systems™ 8064H-500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Suitable for use in ATEX areas
- Push the sampler into the product, rotate the sampler then withdraw
- Deposit the sample into a suitable container
- Place the used sampler into its original plastic bag and dispose
- No cross contaminatin
- No cleaning required
Specifica
Probe | |
Prevents sample from falling out | |
Non-sterile | |
600 mm |
HDPE | |
125 mL | |
Powder Sampling | |
500 mm |
Titolo del prodotto
Sampling Systems™ SteriWare™ Disposable Powder Probe > LengthMetric: 600mm