Learn More
Zeiss™ Stemi 305 Stereo Microscope
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
1960.00€ - 2205.00€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15986182
|
Zeiss™
435063-9020-100 |
Double Spot Illuminator K LED |
2205.00€
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
17332810
|
Zeiss™
435063-9030-100 |
Ring Illuminator K LED |
1960.00€
1 set |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
Description
- This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
- The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
- Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Stereo Microscope |