Learn More
Alpaco™ Morsetti per tubi in acciaio inox
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
Marca: Alpaco™ I-NH 35/10-1636
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Reusable
- Resists pressure
- No lubricant required for assembly
- Smooth clamping to avoid tubing damage
Specifica
10/conf. | |
Fascetta serratubo | |
Acciaio inox | |
30 to 35 mm |