Learn More
Cole-Parmer™ Stainless Steel Female Reducer
Threaded adapters offer a more permanent mounting
Marca: Cole-Parmer™ 31211-10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Threaded fittings
- Sturdy threaded fittings designed for permanent installation
- Use with rigid tubing for high-pressure applications
Specifica
1/4 x 1/8 in. NPT(F) Connection | |
Stainless Steel | |
1/8 in. NPT(F) | |
1/4 x 1/8 in. NPT(F) |
Autoclavable | |
1/4 in. NPT(F) | |
Autoclave, Ethylene Oxide or Gamma | |
Female Reducer |