Learn More
Bochem™ Stainless Steel Beakers
Click to view available options
Capacità (sistema metrico):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Include:
Rim
Rim and Spout
Rim, Handle
Rim, Spout
Rim, Spout, Handle
Descrizione
- 18/10 stainless steel construction.
- Resistant and durable.
- Contact bottom for heating plates and magnetic stirrers.
Specifica
Specifica
Capacità (sistema metrico) | 3000 mL |
Materiale | Stainless Steel (18/10) |
Include | Rim, Spout, Handle |
Tipo | Becher |