Learn More
SPOCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22243-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
SPOCD1 | |
Polyclonal | |
Unconjugated | |
SPOCD1 | |
RP11 84A19.1, SPOC domain containing 1, SPOCD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90853 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZMY3 | |
SPOCD1 | |
SPOCD1 Fusion Protein Ag17734 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Titolo del prodotto
SPOCD1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated