Learn More
BRAND™ Spare Burette Tip
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
Description
Specifications
Specifications
Color | Clear |
Capacity (Metric) | 10 mL |
For Use With (Equipment) | Compact Automatic Burette, Compact Burette |
Material | Borosilicate Glass |
No. per Pack | 1 |
Volume (Metric) Reservoir | 10 mL |
Type | Buret Tip |