Learn More
Sodium 4-hydroxybenzoate, 99+%
Codice prodotto. p-7037778
Click to view available options
Quantità:
100 g
250 g
Dimensione della confezione:
100g
250g
CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 114-63-6
Formula molecolare C7H5NaO3
Molecular Weight (g/mol) 160.10
Numero MDL MFCD00016530
InChI Key ZLVSYODPTJZFMK-UHFFFAOYSA-M
Sinonimo sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1
PubChem CID 16219477
SMILES [Na+].OC1=CC=C(C=C1)C([O-])=O
Specifica
Materiale o nome chimico Sodium 4-hydroxybenzoate
Quantità 100 g
Beilstein 4163553
Informazioni di solubilità Soluble in water.
IUPAC Name sodium;4-hydroxybenzoate
Peso formulazione 160.1
Percent Purity ≥99%