Learn More
Liebherr™ SmartFrost Comfort Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ SmartFrost Comfort Freezers provide a more flexible storage area with greater capacity and improved energy consumption for lower running costs.
Specifica
Colore Bianco
Tensione 220/240 V
Visualizza altre specifiche
Descrizione
Descrizione
SmartFrost
  • Compared to other static freezers, Liebherr’s SmartFrost technology freezes food faster and reduces ice build-up for simpler and quicker defrosting
Automatic SuperFrost function
  • Makes freezing simple and saves energy
  • Quickly reduces the temperature to -32°C and thus creates the cold air required to preserve vital vitamins
  • As soon as the food is completely frozen, the automatic SuperFrost function switches back to normal mode after max. 65 hours, and thus assists in saving electricity
Child lock
  • The child lock is programmed so as to prevent the appliance being inadvertently switched off
  • A symbol in the MagicEye indicates when the child lock is switched on
FrostSafe
  • With the FrostSafe, the extra-high and removable drawers are closed all round
  • The cold cannot dissipate so quickly when the appliance is opened
  • Transparent front of the compartments guarantees an optimum overview of the frozen items
VarioSpace
  • All freezers with NoFrost and SmartFrost feature drawers and intermediate glass shelves underneath them which can be conveniently removed
  • This results in VarioSpace, the practical system for extra storage space so that space can be created quickly even for larger frozen food items
Slimline door handle with integrated opening mechanism
  • The practical door handles with integrated opening mechanism ensure that the appliance doors can be opened with a minimum of effort even if they are opened several times in rapid succession
  • Handle mechanism that assists in pushing the door from the appliance housing
Specifica
Specifica
Bianco
220/240 V