Learn More
Bollé Safety Slam+ Platinum™ CSP Safety Glasses
Descrizione
- Recommended for extremely hot or extremely cold environments
- Comfort, twin material black and orange temples
- All-round vision
- PLATINUM coating
Specifica
Specifica
Certificazione/Conformità | Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T |
Lens Coating | Anti-fog, Scratch-resistant |
Colore telaio | Black, Orange |
Stile stanghette | Twin-material |
Colore stanghette | Black |