Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes
Disposable plastic cassettes hold tissue specimens during processing, embedding and storage
Descrizione
Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents,
- Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
- Cover can be removed and re-inserted easily without danger of specimen loss
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage