Learn More
Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 105-145m2/g
Codice prodotto. p-7058578
Click to view available options
Quantità:
50 g
250 g
1 kg
Dimensione della confezione:
1kg
250g
50g
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 7631-86-9
Formula molecolare O2Si
Molecular Weight (g/mol) 60.08
Numero MDL MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Sinonimo silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
Specifica
Materiale o nome chimico Silicon (IV) oxide
Nota nome Amorphous fumed, surface treated
Colore White
Forma fisica Powder
Quantità 50 g
Formula lineare SiO2
IUPAC Name silanedione
Peso formulazione 60.09
Odore Odorless
Densità 2.2 g/mL