Learn More
Afora™ Short Tube Funnel
Allow liquids to be added to chemical mixtures at a slow pace with the Afora™ Short Tube Funnel.
Brand:  Afora™ 513/08
Product Code. 11976429
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »
Description
Description
  • Weight: 0.01kg
  • Glass 
  • 80mm
Specifications
Specifications
Glass
Weight: 0.01kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions