Learn More
Lenz Laborglasintrumente Separating Funnels, Acc. to Squibb, Amber, with Solid Clear Glass Plug, Without Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 04018037
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- With NS socket and PE stopper
- Without scale, with solid clear glass plug
- With screwthread retaining device
- Material: borosilicate glass 3.3
Specifica
100 mL | |
Borosilicate Glass (3.3) | |
NS 19/26 |