Learn More
Pacific Handy Cutter Safety Cutter 3-in-1 (Cutter, Tape Splitter, Film Cutter)
The 3-in-1 Safety Cutter is a big hit in retail and supermarkets! It has three adjustable blade lengths for box topping, for shallow window cuts or for deep cuts.
366.00€ - 373.00€
Specifica
Product Type | Safety Cutter |
---|---|
Monouso | Reusable |
Sterilità | Non-sterile |
Codice del prodotto | Marca | Colore | Tipo di maniglia | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Colore | Tipo di maniglia | Prezzo | Quantità e disponibilità | ||||
17759037
|
Pacific Handy Cutter
S5L |
Red | Left-Hand |
366.00€
pezzi 12 |
Spedizione stimata: 05-06-2025
Eseguire il login per visualizzare lo stock disponibile |
||||
17769037
|
Pacific Handy Cutter
S5R |
Green | Right-hand |
373.00€
pezzi 12 |
Spedizione stimata: 05-06-2025
Eseguire il login per visualizzare lo stock disponibile |
||||
Descrizione
- The extremely stable metal blade guard protects the user.
- At the same time, the angled guard serves as a cutting guide, e.g. when topping boxes.
- At the end of the handle there is a fold-out, film cutter with concealed blade, which is ideal for foils and strapping.
- Integrated robust tape splitter.
- Spare blades can be comfortably placed under control in the compartment.
- Tool-free blade change.
- With practical eyelet for lanyard fastening or hanging.
- The green model is suitable for right-handed users.
- The red model is suitable for left-handed users.
Specifica
Safety Cutter | |
Reusable | |
Non-sterile |