missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA554179
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: VLESEAGQPQ ALGSSGTCSS LKKQKLRAES DFVKFDTPFL PKPLFFRRAK SSTATHPPGP AVQLNKTPSS SKKVTFGLNR NMTAEFKKTD KSILVSPTGP SRVAFDPEQK PLHGVLKTPT SSPASSPLVA KKPLTT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
RRP1B belongs to the RRP1 family. It may be a novel susceptibility gene for breast cancer progression and metastasis.
Specifica
RRP1B | |
Polyclonal | |
Unconjugated | |
RRP1B | |
2600005C20Rik; D030064A17; KIAA0179; LOW QUALITY PROTEIN: ribosomal RNA processing protein 1 homolog B; mKIAA0179; Nnp1; NNP1L; PPP1R136; protein phosphatase 1, regulatory subunit 136; RGD1305633; ribosomal RNA processing 1 homolog B; ribosomal RNA processing 1 homolog B (S. cerevisiae); ribosomal RNA processing 1B; ribosomal RNA processing protein 1 homolog B; RRP1; RRP1B; RRP1-like protein B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14684 | |
RRP1B | |
Recombinant protein corresponding to Human RRP1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto