Learn More
RIPK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15366-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Serine/threonine/tyrosine kinase that plays an essential role in modulation of innate and adaptive immune responses. Upon stimulation by bacterial peptidoglycans, NOD1 and NOD2 are activated, oligomerize and recruit RIPK2 through CARD-CARD domains. Contributes to the tyrosine phosphorylation of the guanine exchange factor ARHGEF2 through Src tyrosine kinase leading to NF-kappaB activation by NOD2. Once recruited, RIPK2 autophosphorylates and undergoes ′Lys-63′-linked polyubiquitination by E3 ubiquitin ligases XIAP, BIRC2 and BIRC3. The polyubiquitinated protein mediates the recruitment of MAP3K7/TAK1 to IKBKG/NEMO and induces ′Lys-63′-linked polyubiquitination of IKBKG/NEMO and subsequent activation of IKBKB/IKKB. In turn, NF-kappa-B is released from NF-kappa-B inhibitors and translocates into the nucleus where it activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Plays also a role during engagement of the T-cell receptor (TCR) in promoting BCL10 phosphorylation and subsequent NF-kappa-B activation.Specifica
RIPK2 | |
Polyclonal | |
Unconjugated | |
RIPK2 | |
CARD3, CARDIAK, CCK, GIG30, Receptor interacting protein 2, RICK, RIP 2, RIP2, RIPK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192656, 362491, 8767 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43353, P58801 | |
Ripk2 | |
RIPK2 Fusion Protein Ag0909 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Titolo del prodotto
RIPK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated