Learn More
Relative Density Standard 25°C, UKAS ISO 17025 / ISO 17034, 500 mL, nominal Relative Density 25/4 0.8366 at 25 °C, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 Relative Density standards are designed for the calibration or verification of instruments used to measure density and relative density.ISO 17025 / ISO 17034 certification under UKAS ensures full traceability to national standards.
Brand: Paragon Scientific RDEN25-04
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- The Relative density was calculated by dividing the density, as obtained from ASTM D1480, by the density of water at the reference temperature.
- The density of water was taken from the table denoted 'Density of Water' found in ASTM D1480 and confirms to the International Temperature Scale (ITS 90).
- Supplied in tamper evident packaging with a 2 year shelf life
- Manufactured in the United Kingdom
Specifications
500 mL |
Safety and Handling
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Kerosine : distillates, hydrotreated light, White mineral oil, petroleum
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nominal values quoted. Certified Values will vary from batch to batch.