Learn More
Relative Density Standard 15°C, UKAS ISO 17025 / ISO 17034, 500 mL, nominal Relative Density 15/4 0.7186 at 15 °C, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 Relative Density standards are designed for the calibration or verification of instruments used to measure density and relative density.ISO 17025 / ISO 17034 certification under UKAS ensures full traceability to national standards.
Brand:  Paragon Scientific RDEN15-02
Product Code. 16090300
  • 442.00CHF / 500mL
Only null left
Estimated Shipment: 28-07-2025
Add to basket
Description
Description
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • The Relative density was calculated by dividing the density, as obtained from ASTM D1480, by the density of water at the reference temperature.
  • The density of water was taken from the table denoted 'Density of Water' found in ASTM D1480 and confirms to the International Temperature Scale (ITS 90).
  • Supplied in tamper evident packaging with a 2 year shelf life
  • Manufactured in the United Kingdom
Specifications
Specifications
500 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Kerosine : distillates, hydrotreated light, White mineral oil, petroleum
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Nominal values quoted. Certified Values will vary from batch to batch.