Learn More
Ebro Electronic™ AX 400 / RB 1691 B-S Scatola relè
Scatola relè
Marca: Ebro Electronic™ 1340-0157
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Therm. pointed probe (Needle length: 120 mm, dia. 3 mm), cable 1.5m
Specifica
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
For Electronic Contact Thermometer GFX 460 | |
250 mm | |
1.2 kg | |
180 mm |
Single-use | |
180 mm | |
AX 400 | |
Relais Box |